missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide, for analysis, 35 wt.% solution in water, stabilized

Product Code. p-4090151 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
10 L
25 L
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
25 L
25mL
500mL
This item is not returnable. View return policy

Product Code. 10763201

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7722-84-1 | H2O2 | 34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1, 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Boiling Point 108.0°C
Melting Point -33.0°C
Color Yellow through Brown
Physical Form Powder
Quantity 25 mL
Linear Formula H2O2
Fieser 01,457; 02,216; 03,154; 04,253; 05,337; 06,286; 07,174; 08,247; 09,241; 10,201; 11,256; 12,242; 14,176; 15,166; 16,177; 17,145
Merck Index 15, 4835
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and sulfuric ether, insoluble in petroleum ether
Formula Weight 34
Percent Purity 34.5 to 37%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 1.135
Density 1.1350g/mL
Chemical Name or Material Hydrogen peroxide, For analysis, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrogen Peroxide, 35 wt% solution in water, stabilized
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Hydrogen peroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.