Learn More
Hydrogen peroxide, for analysis, 35 wt.% solution in water, stabilized, ACROS Organics™
Brand: Acros Organics 202460010
31.64 EUR valid until 2021-02-28
Use promo code "14802" to get your promotional price.
Code : X2
Additional Details : CAS Number : 7722-84-1 Weight : 1.18816kg Transport : UN number : 2014 Chem class : 5.1 Pack group : II
Packaging | Plastic bottle |
---|---|
Quantity | 1L |
Description
Oxidation reagentChemical Identifiers
7722-84-1 | |
34 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |
H2O2 | |
MFCD00011333 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
CHEBI:16240 | |
OO |
Specifications
Hydrogen peroxide | |
-33.0°C | |
7722-84-1,7732-18-5 | |
H2O2 | |
01,457; 02,216; 03,154; 04,253; 05,337; 06,286; 07,174; 08,247; 09,241; 10,201; 11,256; 12,242; 14,176; 15,166; 16,177; 17,145 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and sulfuric ether, insoluble in petroleum ether | |
OO | |
34 | |
CHEBI:16240 | |
Powder | |
Analysis | |
0.2ppm max. | |
10ppm max. | |
Plastic bottle | |
1.135 | |
108.0°C |
Yellow to Brown | |
For analysis, Stabilized | |
H2O2 | |
MFCD00011333 | |
15, 4835 | |
3ppm max. | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
34 | |
34.5 to 37% | |
2ppm max. | |
1.1350g/mL | |
0.5ppm max. | |
0.025% max. (105°C) | |
about 0.03% stabilizers | |
1L |