missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative, 2.5 to 3.5%, U.S.P., J.T. Baker™

Product Code. 15567984 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
4 L
500mL
This item is not returnable. View return policy

Product Code. 15567984

Brand: Avantor J.T.Baker 218201

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Quantity 500 mL
Formula Weight 34.01
Percent Purity 2.5 to 3.5%
Grade USP
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.00kg
Chemical Name or Material Hydrogen Peroxide, Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.