missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative, 2.5 to 3.5%, U.S.P., J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
4 L
500mL
Specifications
Specifications
| Quantity | 4 L |
| Formula Weight | 34.01 |
| Percent Purity | 2.5 to 3.5% |
| Grade | USP |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 1.00kg |
| Chemical Name or Material | Hydrogen Peroxide, Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative |
Säkerhet och hantering
Produktbeteckning
- Hydrogen peroxide 3.5%
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Hydrogen peroxide
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering