Learn More
Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min
Description
Hydrogen tetrachloroaurate(III) hydrate is used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow to Orange |
| Molecular Formula | AuCl4H |
| Quantity | 5 g |
| UN Number | UN3260 |
| Sensitivity | Hygroscopic; Light sensitive |
| Synonym | Tetrachloroauric(III) acid; Chloroauric acid |
| Solubility Information | Soluble in water. Insoluble in ether. |
| InChI Key | RFWCEGCWRSNAGT-UHFFFAOYSA-K |
| Molecular Weight (g/mol) | 339.77 |
| Formula Weight | 339.79 (Anhydrous) |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.