missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Tetrafluoroborate(1-) (48% solution in H2O), TRC

Product Code. 30389689
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389689 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389689 Supplier Toronto Research Chemicals Supplier No. TRCH71541010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrogen Tetrafluoroborate(1-)
CAS 16872-11-0
Quantity 10 g
Molecular Formula BF4 . H
Synonym Borofluoric Acid, Fluoroboric Acid, Tetrafluoroboric Acid
InChI Formula InChI=1S/BF4/c2-1(3,4)5/q-1/p+1
SMILES F[B-](F)(F)F
IUPAC Name hydron;tetrafluoroborate
Molecular Weight (g/mol) 87.81
Formula Weight 88.0107
Concentration or Composition (by Analyte or Components) 48 %
Show More Show Less
Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.