missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Hydrophilic Dialysis Tubing

Product Code. 11327030
Change view
Click to view available options
MWCO:
1000 kDa
300 kDa
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Unit Size
11327030 300 kDa Each
12787516 1000 kDa Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327030 Supplier Spectrum™ Labs Supplier No. 131450
  • ONLINE EXCLUSIVE PRICE
    866.00 €
    579.00 € / Pack of 1
    Save 287.00 €
    33% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ultra high purity separation dialysis membranes

An ultra high purity separation dialysis membrane. The Spectra/Por™ Cellulose Ester (CE) dialysis membranes offer an excellent Molecular Weight Cut Off (MWCO) selection for separation of ionic species as well as macromolecules. The advance manufacturing process provides CE dialysis membranes with an extremely high purity and features a biological inert surface to yield an optimum sample recovery. Symmetric tubular CE membranes are supplied in form of non-sterile, wet tubular membrane.
Chemical resistance
Variables in temperature, concentrations, durations of exposure and other factors may affect the use of the product. CE membranes are sensitive to organic solvents at concentrations exceeding 5% in water. Strong polar solvents such as acetone, Methyl Ethyl Ketone (MEK) or Dioxane will irreparably damage the membranes. Lower alcohols such as Methanol, and Isopropanol can be used with CE membranes at short exposure or low concentrations.
  • Hydrophilic and uniform molecular porous dialysis membranes
  • Nine MWCOs: 100D to 500D, 500D to 1,000D, 3, 5kD to 5kD, 8kD to 10kD, 20kD, 50kD, 100kD, 300kD and 1,000kD
  • Four flat widths: 10mm, 16mm, 24mm, 33mm
  • No special chemical pre-cleaning required
  • Non-sterile Biotech tubular CE membranes
  • Supplied wet in 0.1% Sodium Azide
  • Supplied in 10m length (wet)

Specifications

Product Type Dialysis Tubing
MWCO 300 kDa
Diameter (Metric) 10 mm
Length (Metric) 10 m
Capacity (Metric) 0.79 mL/cm
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.