missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Hydrophobic Coverslips
Description
- The coverslips should be used to conserve reagents when larger sample areas are being labeled.
- Unlike glass coverslips that may not wet evenly and may be too heavy to allow sufficient reagent to contact the sample, these coverslips float easily on small volumes of labeling reagents.
- Unlike wax film or other commercially available coverslips, these coverslips are rigid, preventing uneven distribution of labeling solutions.
- The coverslips are packaged in rows of 5 with a protective film coating.
- Dimensions: 22 x 40mm
- Thickness 0.25 mm thick (each)
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70°C |
| Quantity | 100 Each |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?