missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Cyclophosphamide Semicarbazone (80%), TRC

Product Code. 30137858
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137858 25 mg 25mg
30011596 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137858 Supplier Toronto Research Chemicals Supplier No. TRCH92630525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53948-46-2
Molecular Formula C8H18Cl2N5O3P
Molecular Weight (g/mol) 334.14
Synonym N,N-bis(2-chloroethyl)phosphorodiamidic Acid 3-[2-(Aminocarbonyl)hydrazinylidene]propyl Ester, NSC 319519, Aldophosphamide Semicarbazone;
SMILES O=P(N(CCCl)CCCl)(N)OCC/C=N/NC(N)=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H18Cl2N5O3P/c9-2-5-15(6-3-10)19(12,17)18-7-1-4-13-14-8(11)16/h4H,1-3,5-7H2,(H2,12,17)(H3,11,14,16)/b13-4+
Formula Weight 333.0524
Chemical Name or Material Hydroxy Cyclophosphamide Semicarbazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.