missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Dimetridazole, TRC

Product Code. 17992475
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992475 50 mg 50mg
17991365 5 mg 5mg
17963185 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992475 Supplier Toronto Research Chemicals Supplier No. TRCH82524550MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 936-05-0
Molecular Formula C5 H7 N3 O3
Molecular Weight (g/mol) 157.127
Synonym (1-Methyl-5-nitroimidazol-2-yl)methanol, Imidazole-2-methanol, 1-methyl-5-nitro- (7CI,8CI), 1-Methyl-5-nitro-1H-imidazole-2-methanol, (1-Methyl-5-nitro-1H-imidazol-2-yl)-methanol, 1-Methyl-2-hydroxymethyl-5-nitroimidazole, 1-Methyl-5-nitro-2-imidazolemethanol, 2-Hydroxymethyl-1-methyl-5-nitroimidazole, Hydroxydimetridazole
IUPAC Name (1-methyl-5-nitroimidazol-2-yl)methanol
SMILES Cn1c(CO)ncc1[N+](=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H7N3O3/c1-7-4(3-9)6-2-5(7)8(10)11/h2,9H,3H2,1H3
Formula Weight 157.049
Percent Purity >95%
Chemical Name or Material Dimetridazole-2-hydroxy
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.