missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Ipronidazole, TRC

Product Code. 30009015
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009015 25 mg 25mg
17921925 10 mg 10mg
17973495 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009015 Supplier Toronto Research Chemicals Supplier No. TRCH94353525MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35175-14-5
Molecular Formula C7 H11 N3 O3
Molecular Weight (g/mol) 185.18
Synonym α,α,1-Trimethyl-5-nitro-1H-imidazole-2-methanol, 1,α,α-Trimethyl-5-nitroimidiazole-2-methanol, 1-Methyl-2-(2'-hydroxyisopropyl)-5-nitroimidazole, 2-(1-Hydroxy-1-methylethyl)-1-methyl-5-nitroimidazole, Hydroxyipronidazole, Ipronidazole-OH, L 12407, Ipronidazole-hydroxy
IUPAC Name 2-(1-methyl-5-nitroimidazol-2-yl)propan-2-ol
SMILES Cn1c(cnc1C(C)(C)O)[N+](=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C7H11N3O3/c1-7(2,11)6-8-4-5(9(6)3)10(12)13/h4,11H,1-3H3
Formula Weight 185.08
Percent Purity >95%
Chemical Name or Material Ipronidazole-hydroxy
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.