missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Urea, TRC

Product Code. 30040830
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040830 10 mg 10mg
18003120 50 mg 50mg
18090820 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040830 Supplier Toronto Research Chemicals Supplier No. TRCH99100010MG
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-07-1
Molecular Formula C H4 N2 O2
Molecular Weight (g/mol) 76.05
Synonym N-Hydroxyurea, Biosupressin, Carbamohydroxamic Acid, Carbamoyl Oxime, Cytodrox, Droxia, HU, Hidrix, Hydrea, Hydreia, Hydroxycarbamide, Hydroxycarbamine, N-(Aminocarbonyl)hydroxylamine, Hydroxylurea, Hydura, Hydurea, Litaler, Litalir, N-Carbamoylhydroxylamine, NCI C04831, NSC 32065, Oxyurea, SK 22591, SQ 1089
IUPAC Name hydroxyurea
SMILES NC(=O)NO

Specifications

Quantity 10 mg
InChI Formula InChI=1S/CH4N2O2/c2-1(4)3-5/h5H,(H3,2,3,4)
Formula Weight 76.0273
Chemical Name or Material Hydroxycarbamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.