missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyacetone, 95%

Product Code. 11378106
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378106 500 g 500g
11368106 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378106 Supplier Thermo Scientific Alfa Aesar Supplier No. L15008.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 116-09-6 | C3H6O2 | 74.079 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Hydroxyacetone is used as a reagent in organic chemical reactions. It also serves as a component for Mannich reaction and aldol reactions. It is also used in the syntheses of 2-oxo-propionaldehyde, imidazoles, polyols, acrolein, dyes and skin tanning agents. It yields (R)-1,2-propanediol upon reduction of hydroxyacetone in the presence of a microbial cell catalyst.

Solubility
Miscible with water, alcohol and ether.

Notes
Store in cool place. Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-09-6
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00004669
InChI Key XLSMFKSTNGKWQX-UHFFFAOYSA-N
Synonym hydroxyacetone, acetol, acetone alcohol, 1-hydroxy-2-propanone, 2-propanone, 1-hydroxy, 1-hydroxyacetone, methanol, acetyl, acetylmethanol, acetylcarbinol, 2-oxopropanol
PubChem CID 8299
ChEBI CHEBI:27957
IUPAC Name 1-hydroxypropan-2-one
SMILES CC(=O)CO

Specifications

Melting Point -17°C
Density 1.08
pH 3.4
Boiling Point 145°C to 146°C
Flash Point 56°C (132°F)
Odor Characteristic
Refractive Index 1.425
Quantity 500 g
UN Number UN1224
Beilstein 605368
Sensitivity Hygroscopic
Merck Index 14,65
Solubility Information Miscible with water,alcohol and ether.
Formula Weight 74.08
Percent Purity 95%
Chemical Name or Material Hydroxyacetone
Show More Show Less
compliance-icons
Product Identifier
  • Hydroxyacetone
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: Hydroxyacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.