missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyacetone, Technical

Product Code. 10021504 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10021504 500 g Glass bottle 500g
10306494 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10021504 Supplier Thermo Scientific Acros Supplier No. 160345000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 116-09-6 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-09-6
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00004669
InChI Key XLSMFKSTNGKWQX-UHFFFAOYSA-N
Synonym hydroxyacetone, acetol, acetone alcohol, 1-hydroxy-2-propanone, 2-propanone, 1-hydroxy, 1-hydroxyacetone, methanol, acetyl, acetylmethanol, acetylcarbinol, 2-oxopropanol
PubChem CID 8299
ChEBI CHEBI:27957
IUPAC Name 1-hydroxypropan-2-one
SMILES CC(=O)CO

Specifications

Melting Point -17.0°C
Density 1.0800g/mL
Boiling Point 145.0°C to 146.0°C
Flash Point 56°C
Packaging Glass bottle
Linear Formula CH3COCH2OH
Refractive Index 1.4315
Quantity 500 g
UN Number 1224
Beilstein 01, 821
Merck Index 15, 64
Specific Gravity 1.08
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Viscosity 1.593 mPa.s (48°C)
Formula Weight 74.08
Grade Technical
Chemical Name or Material Hydroxyacetone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.