Learn More
Tocris Bioscience™ Hydroxychloroquine sulfate
Autophagy inhibitor; also TLR9 inhibitor
Brand:  Tocris Bioscience™ 5648/10
Encompass
Product Code. 15796745
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Autophagy inhibitor. Inhibits growth and induces apoptosis of renal cancer cells in vitro. Also inhibits TLR9. Additionally inhibits PRC2 by inhibiting the allosteric binding of PRC2 to EED (embryonic ectoderm development) within the H3K27me3-binding pocket, and reduces H3K27me3 levels in multiple myeloma cells in vitro.
Hydroxychloroquine sulfate is an autophagy inhibitor. Also inhibits TLR9. Inhibits growth and induces apoptosis of renal cancer cells in vitro. Additionally inhibits PRC2 by inhibiting the allosteric binding of PRC2 to EED (embryonic ectoderm development) within the H3K27me3-binding pocket, and reduces H3K27me3 levels in multiple myeloma cells in vitro. Inhibits SARS-CoV-2 viral infection, in vitro (EC50 values in μM range and dependent on viral RNA copy number). Also antimalarial and immunomodulator.
Tocris products are for biomedical research use only. They are not intended for human or veterinary use.
Specifications
Specifications
2-[[4-[(7-Chloro-4-quinolinyl)amino]pentyl]ethylamino]ethanol sulfate
747-36-4
10 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.