Learn More
Tocris Bioscience™ Hydroxychloroquine sulfate
GREENER_CHOICE
Autophagy inhibitor; also TLR9 inhibitor
Brand:  Tocris Bioscience™ 5648/50
Encompass
Product Code. 15706755
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Autophagy inhibitor. Inhibits growth and induces apoptosis of renal cancer cells in vitro. Also inhibits TLR9. Additionally inhibits PRC2 by inhibiting the allosteric binding of PRC2 to EED (embryonic ectoderm development) within the H3K27me3-binding pocket, and reduces H3K27me3 levels in multiple myeloma cells in vitro.
Hydroxychloroquine sulfate is an autophagy inhibitor. Also inhibits TLR9. Inhibits growth and induces apoptosis of renal cancer cells in vitro. Additionally inhibits PRC2 by inhibiting the allosteric binding of PRC2 to EED (embryonic ectoderm development) within the H3K27me3-binding pocket, and reduces H3K27me3 levels in multiple myeloma cells in vitro. Inhibits SARS-CoV-2 viral infection, in vitro (EC50 values in μM range and dependent on viral RNA copy number). Also antimalarial and immunomodulator.
Tocris products are for biomedical research use only. They are not intended for human or veterinary use.
Specifications
Specifications
2-[[4-[(7-Chloro-4-quinolinyl)amino]pentyl]ethylamino]ethanol sulfate
747-36-4
50 mg
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Product Certifications
Promotions
Promotions
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.