missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Hydroxychloroquine sulfate
GREENER_CHOICE

Product Code. 18750298
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18750298 10 mg 10mg
18720308 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18750298 Supplier Tocris Bioscience™ Supplier No. 5648/10
  • ONLINE EXCLUSIVE PRICE
    153.00 €
    143.85 € / 10mg
    Save 9.15 €
    5% Off
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Autophagy inhibitor; also TLR9 inhibitor

Autophagy inhibitor. Inhibits growth and induces apoptosis of renal cancer cells in vitro. Also inhibits TLR9. Additionally inhibits PRC2 by inhibiting the allosteric binding of PRC2 to EED (embryonic ectoderm development) within the H3K27me3-binding pocket, and reduces H3K27me3 levels in multiple myeloma cells in vitro.

Hydroxychloroquine sulfate is an autophagy inhibitor. Also inhibits TLR9. Inhibits growth and induces apoptosis of renal cancer cells in vitro. Additionally inhibits PRC2 by inhibiting the allosteric binding of PRC2 to EED (embryonic ectoderm development) within the H3K27me3-binding pocket, and reduces H3K27me3 levels in multiple myeloma cells in vitro. Inhibits SARS-CoV-2 viral infection, in vitro (EC50 values in μM range and dependent on viral RNA copy number). Hydroxychloroquine improves efficacy of adeno-associated viral gene transduction in vivo and in vitro. Also antimalarial and immunomodulator. Tocris products are for biomedical research use only. They are not intended for human or veterinary use.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[[4-[(7-Chloro-4-quinolinyl)amino]pentyl]ethylamino]ethanol sulfate
CAS 747-36-4
Quantity 10 mg
Target Autophagy-related Compounds
Molecular Formula C18H28ClN3O5S
Formula Weight 433.95
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.