missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxylamine-d3 DCl, CDN
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Hydroxylamine-d3 Hydrochloride-d |
| CAS | 15588-23-5 |
| Molecular Formula | Cl D . D3 N O |
| Quantity | 5 g |
| Synonym | Hydrochloric acid-d, compd. with hydroxylamine-d3 (1:1) (9 CI), Hydroxyl-d-amine-d2, hydrochloride-d (6 CI), Hydroxylamine-d3, hydrochloride-d (7 CI, 9 CI), Hydroxylamine-d3 DCl, Trideuteriohydroxylamine deuterochloride, Hydroxylamine hydrochloride D4 |
| InChI Formula | InChI=1 S/ClHH3NO/c;1-2/h1H;2 H,1H2/i;1D2,2 D/hD |
| SMILES | [2 H]Cl.[2 H]ON([2 H])[2 H] |
| Molecular Weight (g/mol) | 73.516 |
| Formula Weight | 73.023 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction