missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, 96% min., MP Biomedicals

Product Code. 11384469 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384469 1 kg 1kg
11374469 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384469 Supplier MP Biomedicals Supplier No. 0210199791
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Slowly decomposes when moist

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine hydrochloride
SMILES Cl.NO

Specifications

Quantity 1 kg
Assay Percent Range 96% min.
Formula Weight 69.5
Chemical Name or Material Hydroxylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.