missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine hydrochloride, 97%

Product Code. 10245840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10245840 1 kg 1kg
10124380 100 g 100g
10184420 500 g 500g
15204664 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10245840 Supplier Thermo Scientific Acros Supplier No. 170360010
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hydroxylamine hydrochloride, 97%, Quantity: 1kg, CAS: 5470-11-1, White, Melting Point: 155 deg.C to 158 deg.C, Molecular Formula: H3NO.HCl, Molecular Weight: 69.49, Packaging: Plastic bottle, Percent Purity: 97% | CAS: 5470-11-1 | ClH4NO | 69.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine;hydrochloride
SMILES Cl.NO

Specifications

Melting Point 155°C to 158°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 1 kg
Assay Percent Range 97%
Linear Formula H2NOH·HCl
Fieser 01,478; 02,217; 07,176; 09,245
Merck Index 15,4867
Solubility Information Solubility in water: 560g/L (20°C). Other solubilities: soluble in alcohol,methanol,glycerol,propylene glycol,insoluble in cold ether
Formula Weight 69.49
Percent Purity 97%
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Hydroxylamine hydrochloride, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.