missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, ACS Reagent Grade, Scharlab

Product Code. 11314184
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314184 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11314184 Supplier Scharlab Supplier No. HI02150250
  • ONLINE EXCLUSIVE PRICE
    248.97 €
    245.02 € / 250g
    Save 3.95 €
    1% Off
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in analytical chemistry, as a strong reducing agent and in synthesis of organic products. Scharlab Hydroxylamine Hydrochloride, ACS Reagent Grade is also used for the synthesis of oximes, for the separation and identification of tellurium and selenium, and for the determination of gold.

  • Synonyms: Hydroxylammonium chloride
  • Molecular Formula: NH2OH • HCl
  • Molar Mass: 69.49g/mol
  • CAS: 5470-11-1
  • EIN ECS No.: 226-798-2

Specifications

Melting Point 159°C
CAS 5470-11-1
pH 2.5–4.0 (5%, water)
Quantity 250 g
Molecular Formula ClH4NO
MDL Number MFCD00051089
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
SMILES Cl.NO
IUPAC Name hydroxylamine hydrochloride
Molecular Weight (g/mol) 69.49
PubChem CID 443297
ChEBI CHEBI:5807
Formula Weight 69.49g/mol
Grade ACS Reagent
Identification Passes Test (IR spectrum)
Loss on Drying 0.1% max (105°C)
Residue after Ignition 0.01% max
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydroxylamine hydrochloride, reagent grade, ACS, ISO
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Substances and mixtures corrosive to metals Category 1
  • ACUTE AQUATIC Acute 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302+H312-Harmful if swallowed or in contact with skin
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.