missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine sulfate, 99%

Product Code. 11389349
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
Unit Size:
100g
25g
2kg
500g
This item is not returnable. View return policy

Product Code. 11389349

Brand: Thermo Scientific Alfa Aesar 088944.A3

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10039-54-0 | H3NO·0.5H2SO4

Hydroxylammonium sulfate is used to produce hydroxylamine-O-sulfonic acid from chlorosulfuric acid. It is widely used in organic synthesis for the conversion of aldehydes and ketones to oximes, carboxylic acids and their derivatives (e.g. esters) to hydroxamic acids, isocyanates to N-hydroxyureas and nitriles to amidoximes. It acts as a radical scavenger which is used to terminate the radical polymerization reactions. It finds application as an antioxidant in natural rubber. It is also used in photography as a stabilizer for color developers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10039-54-0
Molecular Formula H3NO·0.5H2SO4
MDL Number MFCD00044869

Specifications

Melting Point ∼170°C (decomposition)
Color White
pH 3.6
Physical Form Crystalline
Quantity 2 kg
Linear Formula (NH2OH)2·H2SO4
UN Number UN2865
Merck Index 14,4828
Solubility Information Soluble in water.
Formula Weight 164.14
Percent Purity 99%
Odor Odorless
Density 1.88
Chemical Name or Material Hydroxylamine sulfate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.