missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Sulfate, TRC

Product Code. 30391492
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30391492 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30391492

Brand: Toronto Research Chemicals TRCH94383050G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Hydroxylamine Sulfate
CAS 10039-54-0
Molecular Formula H8N2O6S
Quantity 50 g
Synonym Bis(hydroxylamine) Sulfate, Bis(hydroxylammonium) Sulfate, Di(hydroxylamine) Sulfate, Envision MLB 2030, Hydroxyl Ammonium Sulfate ((HONH3)2SO4), Hydroxylamine Neutral Sulfate, Hydroxylammonium Sulfate, Lanasane LAB, Oxammonium Sulfate
InChI Formula InChI=1S/2H4NO.H2O4S/c2*1-2;1-5(2,3)4/h2*2H,1H3;(H2,1,2,3,4)/q2*+1;/p-2
SMILES NO.NO.OS(=O)(=O)O
IUPAC Name hydroxylamine;sulfuric acid
Molecular Weight (g/mol) 164.14
Formula Weight 164.0103
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.