missing translation for 'onlineSavingsMsg'
Learn More

Hydroxynaphthol Blue, Indicator for Metal Titration, Solstice

Product Code. 15634110
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass Bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15634110 50 g Glass Bottle 50g
15624110 10 g Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15634110 Supplier Honeywell-Fluka Supplier No. 3393650G
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Indicator for metal titration

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63451-35-4
Molecular Formula C20H11N2Na3O11S3
Molecular Weight (g/mol) 620.46
MDL Number MFCD00036393
InChI Key JQECMNGWLIFQLL-UHFFFAOYSA-K
PubChem CID 44134769
IUPAC Name trisodium;3-hydroxy-4-[2-(2-oxo-4-sulfonatonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+]

Specifications

UN Number NONH for all modes of transport
Infrared Spectrum Conforms to structure
Formula Weight 620.47g/mol
Packaging Glass Bottle
Quantity 50 g
Physical Form Liquid
Chemical Name or Material Hydroxynaphthol blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.