missing translation for 'onlineSavingsMsg'
Learn More

Hydroxynapthol Blue (Technical Grade), TRC

Product Code. 30404263
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404263 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404263 Supplier Toronto Research Chemicals Supplier No. TRCH9502601G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroxynapthol Blue
CAS 63451-35-4
Molecular Formula C20 H11 N2 O11 S3 . 3 Na
Quantity 1 g
Synonym 3-Hydroxy-4-[(2-hydroxy-4-sulfo-1-naphthalenyl)azo]-2,7-naphthalenedisulfonic Acid Trisodium Salt
InChI Formula InChI=1S/C20H14N2O11S3.3Na/c23-15-9-16(35(28,29)30)13-3-1-2-4-14(13)18(15)21-22-19-12-6-5-11(34(25,26)27)7-10(12)8-17(20(19)24)36(31,32)33;;;/h1-9,23-24H,(H,25,26,27)(H,28,29,30)(H,31,32,33);;;/q;3*+1/p-3/b22-21+;;;
SMILES [Na+].[Na+].[Na+].Oc1cc(c2ccccc2c1N=Nc3c(O)c(cc4cc(ccc34)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC Name trisodium;3-hydroxy-4-[(E)-(2-hydroxy-4-sulfonatonaphthalen-1-yl)diazenyl]naphthalene-2,7-disulfonate
Molecular Weight (g/mol) 620.47
Formula Weight 619.9218
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.