missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Hydroxypropyl cellulose, Average M.W. 100.000

Product Code. 15129223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
250g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15129223 5 kg Plastic drum 5kg
10723191 100 g Glass bottle 100g
10477380 250 g Glass bottle 250g
10667753 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15129223 Supplier Thermo Scientific Chemicals Supplier No. 184880051
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9004-64-2 | (C24H44O16)n

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-64-2
Molecular Formula (C24H44O16)n
MDL Number MFCD00132688
Synonym 4-1-aminopropyl-n,n,3-trimethylaniline, 4-dimethylamino-alpha,2-dimethylphenethylamine, benzeneethanamine, 4-dimethylamino-alpha,2-dimethyl, amiflamin, 55875-51-9 di-hydrochloride, hydroxypropyl cellulose, m.w. 100,000, 4-1-aminopropyl-n,n,3-trimethyl-aniline, hydroxypropyl cellulose 6-10 mpa?s in water at 20 ?c, hydroxypropyl cellulose, 3-6 mpa?s in water at 20 ?c

Specifications

Color White to Cream
pH 5 to 8.5 (1% soln.)
Physical Form Granules or Crystalline Powder
Quantity 5 kg
Formula Weight 100.000 (Average )
Infrared Spectrum Authentic
Loss on Drying 5% max. (105°C)
Packaging Plastic drum
Viscosity 75-150 mPa.s 5% aq.sol.
Chemical Name or Material Hydroxypropyl cellulose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.