missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl-beta-cyclodextrin, 97%

Product Code. 10175600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10175600 100 g Glass Bottle 100g
10388870 25 g Glass Bottle 25g
10702164 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10175600 Supplier Thermo Scientific Acros Supplier No. 297561000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 128446-35-5 | C44H75O36 | 1180.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128446-35-5
Molecular Formula C44H75O36
Molecular Weight (g/mol) 1180.05
MDL Number MFCD16621721

Specifications

Melting Point 278.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 2% max. Residual beta-cyclodextrin (On dry substance) (HPLC)
Packaging Glass Bottle
Quantity 100 g
Solubility Information Solubility in water: soluble. Other solubilities: methanol: >330g/l (25°C),chloroform: < 10g/l (25°C),dmso: >330g/l (25°C)
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Hydroxypropyl-β-cyclodextrin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.