missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl Nortadalafil, TRC

Product Code. 30403650
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403650 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403650 Supplier Toronto Research Chemicals Supplier No. TRCH99635050MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroxypropyl Nortadalafil
CAS 2520114-73-0
Molecular Formula C24 H23 N3 O5
Quantity 50 mg
Synonym (6S,12aS)-6-(benzo[d][1,3]dioxol-5-yl)-2-(3-hydroxypropyl)-2,3,12,12a-tetrahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4(6H,7H)-dione
InChI Formula InChI=1S/C24H23N3O5/c28-9-3-8-26-12-21(29)27-18(24(26)30)11-16-15-4-1-2-5-17(15)25-22(16)23(27)14-6-7-19-20(10-14)32-13-31-19/h1-2,4-7,10,18,23,25,28H,3,8-9,11-13H2/t18-,23-/m1/s1
SMILES OCCCN1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O
IUPAC Name (2R,8R)-2-(1,3-benzodioxol-5-yl)-6-(3-hydroxypropyl)-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione
Molecular Weight (g/mol) 433.46
Formula Weight 433.1638
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.