missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyurea, 98%

Product Code. 10646162 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10646162 25 g Glass bottle 25g
10795991 5 g Glass bottle 5g
10502164 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10646162 Supplier Thermo Scientific Acros Supplier No. 151680250
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hydroxyurea, 98%, CAS # 127-07-1, is a compound with antineoplastic, immunomodulatory, radical scavenger, and antimitotic activities. It shows inhibitory activity against nucleic acid synthesis. | CAS: 127-07-1 | CH4N2O2 | 76.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Hydroxyurea is a member of the class of urea that shows antineoplastic, immunomodulatory, radical scavenger, and antimitotic activities.

• It has inhibitory activity against nucleic acid synthesis.

• It also inhibits ribonucleoside diphosphate reductase, an enzyme necessary to convert ribonucleoside diphosphate into deoxyribonucleoside diphosphate, preventing cells from leaving the G1/S phase of the cell cycle

Applications

• Hydroxyurea has an anti-retroviral activity, inhibiting human immunodeficiency virus type 1 (HIV-1) DNA synthesis in peripheral blood lymphocytes, decreasing the amount of intracellular deoxynucleotides

• This compound also has a radio sensitizing activity, maintaining cells in the radiation-sensitive G1 cell cycle phase and, consequently, interfering with DNA repair

• It has been used for cell cycle synchronization in human breast carcinoma cells

• This compound presents inhibitory activity in cell adhesion and motility assay in ccRCC Caki (HTB-47) and CD9 knockdown cells

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-07-1
Molecular Formula CH4N2O2
Molecular Weight (g/mol) 76.04
MDL Number MFCD00007943
InChI Key VSNHCAURESNICA-UHFFFAOYSA-N
Synonym hydroxycarbamide, n-hydroxyurea, hydrea, 1-hydroxyurea, oxyurea, carbamoyl oxime, biosupressin, hydroxycarbamine, onco-carbide, carbamohydroxamic acid
PubChem CID 3657
ChEBI CHEBI:44423
IUPAC Name hydroxyurea
SMILES C(=O)(N)NO

Specifications

Melting Point 142°C to 146°C
Color White
Loss on Drying 1% max. (1 g, 60°C) (< 0.7 kPa)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (on dried substance) (Iodometry)
Packaging Glass bottle
Linear Formula HONHCONH2
Quantity 25 g
Beilstein 03, 95
Merck Index 15, 4888
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in hot alcohol
Formula Weight 76.04
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Hydroxyurea
Show More Show Less
compliance-icons
Product Identifier
  • Hydroxyurea
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1A
Hazard Statement
  • H360D-May damage the unborn child.
  • H340-May cause genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.