Learn More
Ansell HyFle™ 11-755 Cut-resistant Gloves
Product Code. 18804417 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
12
Unit Size:
12 pairs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
18804417 7 12 pairs
18814417 8 12 pairs
18844417 12 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18804417 Supplier Ansell Supplier No. 11755070
  • ONLINE EXCLUSIVE PRICE
    338.80 €
    219.94 € / 12 pairs
    Save 118.86 €
    35% Off
Estimated Shipment: 06-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ultra-light ANSI/ISEA 105-2024 CUT A5 and EN ISO E cut-resistant gloves with touchscreen compatibility.
  • Heightened defenses: Designed with INTERCEPT™ Cut Resistance Technology and meeting ANSI/ISEA 105-2024 CUT A5 and EN ISO E standards, HyFlex™ 11-755 industrial gloves protect against lacerations and help prevent common workplace injuries
  • Enhanced tactile sensitivity: Palm-dipped, with nylon liner, these PU-coated gloves benefit from heightened tactile sensitivity and touchscreen compatibility
  • Elevated comfort: Lightweight and fitted to natural hand contours, HyFlex™ 11-755 PPE gloves cause minimal hand fatigue
  • Improved dexterity: These polyurethane gloves' ultralight design also increases flexibility and ease of use
  • Certified skin-friendliness: HyFlex™ 11-755 industrial working gloves are Dermatest™-approved for skin-friendliness
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Polyurethane
Color Gray
Cut Resistance Rating Level E
Hand Specific
Latex Free Yes
Material HPPE, Tungsten, Spandex, Polyester, Polyurethane
Product Type Industrial Gloves
Size 7