missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-605 Light Duty Nylon Gloves with Polyurethane Coated Fingertips

Product Code. 12600722
Change view
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12600722 10 12 pairs
12630722 8 12 pairs
12640722 9 12 pairs
12610722 6 12 pairs
12620722 7 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12600722 Supplier Ansell Supplier No. 11605100
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Industrial gloves designed for light and delicate applications in dry environments that require high levels of dexterity.

  • Nylon gloves are comfortable and flexible
  • Polyurethane coating on the fingertips enhances tactile sensitivity
  • Designed for light and delicate applications that require high levels of dexterity
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Fingertips
Coating Material Polyurethane
Color White, Yellow
Cut Resistance Rating ANSI A1
Exterior Finish Fingertip Textured
Hand Specific
Latex Free Yes
Material Nylon
Product Type Dip-coated Gloves
Size 10
EN388 Standard 003XA
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 11-605
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.