Learn More
Ansell™ HyFlex™ 11-616 Series Black and Green/Blue Polyurethane Light Weight Gloves
Product Code. 12650722
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12650722 10 12 pairs
12660722 11 12 pairs
12670722 6 12 pairs
12690722 8 12 pairs
12680722 7 12 pairs
12600732 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12650722 Supplier Ansell™ Supplier No. 11616100
Only null left
Add to basket
Add to basket
Designed for a wide range of lightweight applications in dry or slightly oily environments. Offers outstanding comfort, fit and sensitivity for the most delicate handling.
  • 18-gauge knitting technology allows this glove to be ultra-thin yet tough
  • Ideal for workers who need a higher level of fit, touch and precision
  • Ideal for applications where cotton gloves or no gloves at all are used despite the risk of scrapes, blisters and minor injuries.
TRUSTED_SUSTAINABILITY
Specifications
Back Material Polyurethane
Certifications/Compliance CE Mark, EN 388 : 3121
Cuff Material Polyurethane
Coating Material Polyurethane
Color Blue
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Nylon
Product Type Lightweight Gloves
Size 10
Glove Cuff Style Knit
Liner Material Nylon
Lining Lined
Palm Type Coated-palm
Quantity 144 Pairs
Series 11-616