Learn More
Ansell™ HyFlex™ 11-619 Series Knitted Polyurethane Light Gloves
Almost a second skin, this very thin glove offers outstanding comfort, fit and sensitivity for the most delicate handling. Ansell HyFlex™ 11-619 Series Knitted Polyurethane Light Gloves feature 18-gauge knitting technology which allows the glove to be ultra-thin yet tough.
Brand:  Ansell™ 11619090
Code : NEW
Additional Details : Weight : 0.29700kg
 View more versions of this product
Product Code. 12660732
  • 9.05 € / 12 pairs
In Stock
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Designed for a wide range of lightweight applications in dry or slightly oily environments
  • Ideal for workers who need a higher level of fit, touch and precision than that offered by their current gloves
  • Also ideal for applications where cotton gloves or no gloves at all are used despite the risk of scrapes, blisters and minor injuries
  • Ideal for final assembly, airco, electrical engines, wire harness assembly, adjusters work
Specifications
Specifications
Polyurethane
Polyurethane
Lightweight Gloves
11-619
Nylon
144 Pairs
CE Mark, EN 388 : 2121
Polyurethane
9
Knit
Coated-palm
11-619