Learn More
Ansell HyFlex™ Medium-Duty Mechanical Protection Cut-Resistant Gloves
Product Code. 12690732
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
144 pairs
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12690732 6 12 pairs
12600742 7 12 pairs
12610742 8 12 pairs
12620742 9 12 pairs
12900055 9 144 pairs
12670732 10 12 pairs
12680732 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12690732 Supplier Ansell Supplier No. 11624060
In Stock
Add to basket
Edge
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Designed for secure grip on dry to slightly oily pieces in a wide range of environments. Offers a combination of comfort, softness, coolness and cut protection.
  • Constructed of DSM Dyneema™, Spandex™ and nylon
  • Ergnomic design promotes wearer comfort
  • Special formulation of polyurethane coating for flexibility and fit provides sure grip with palm-coated finish
  • Can be washed and reused without any loss of cut protection
  • Strong cut resistance
  • Knitwrist cuff
  • Black and gray color
Compliance
ANSI™ Level 2 cut protection.
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 5
Certifications/Compliance (Europe) CE, EN 420:2003 + A1:2009; Category II
Cuff Material Knit
Coating Material Polyurethane
Color Black, White
Cut Resistance Rating ANSI A2
Product Type Cut-resistant Gloves
Size 6
EN388 Standard EN 388:4342
For Use With (Application) Glass manufacturing, assembly, plastics injection and moulding, automotive, handling of sharp edged objects, preassembling, cutting of small dry or lightly oiled parts, maintenance
Glove Cuff Style Knit
Liner Material Lycra™
Model 11-624_6
Series HyFlex™ 11-624