missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-728 Polyurethane-Coated HPPE-Lined Gloves with Intercept™ Technology

Product Code. 16629083
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16629083

Brand: Ansell 11728090

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Gloves combine comfort and dexterity with Intercept™ Technology to provide a high level of cut resistance. Polyurethane coating enhances grip capabilities in dry, as well as slightly oily environments.

  • A combination of comfort, sure handling, and cut resistance
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Ideal for safe use in controlling, and handling/assembly applications that involve contact with sharp pieces
  • Designed for use in dry as well as slightly oily environments
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free No
Material Intercept Technology Yarn
Product Type Cut-resistant Gloves
Size 9
EN388 Standard 4342B
For Use With (Application) Glass Repair, Metal Handling, and Press Injection Molding
Glove Cuff Style Knit
Liner Material HPPE
Series 11-728
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.