missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-730 Polyurethane-Coated Gloves with Intercept™ Technology

Product Code. 16659083
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16659083 6 12 pairs
16669083 7 12 pairs
16679083 8 12 pairs
16689083 9 12 pairs
16699083 10 12 pairs
16609093 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16659083 Supplier Ansell Supplier No. 11730060

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gloves combine comfort and dexterity with Intercept™ Technology to provide a high level of cut resistance. Polyurethane coating enhances grip capabilities in dry, as well as slightly oily environments.

  • A combination of comfort, sure handling, and cut resistance
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Ideal for safe use in controlling, and handling/assembly applications that involve contact with sharp pieces
  • Designed for use in dry as well as slightly oily environments
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 407:2004, CE 0493, EN 420:2003 + A1:2009
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A5
Hand Specific
Latex Free Yes
Material Intercept Technology Yarn
Product Type Cut- and Abrasion-resistant Glove
Size 6
EN388 Standard 4X43D
For Use With (Application) Metalwork, Parts Assembly, and Press Injection Molding
Gender Unisex
Glove Cuff Style Knit
Grip Design Rough
Lining Lined
Puncture Rating ANSI/ISEA 105-16 Level 3
Series 11-730
Thumb Style Wing
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.