missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-738 Intercept™ Cut-Resistant Gloves with Targeted Reinforcement

Product Code. 16029661 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16029661 9 12 pairs
16099651 6 12 pairs
16009661 7 12 pairs
16019661 8 12 pairs
16039661 10 12 pairs
16049661 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16029661 Supplier Ansell Supplier No. 11738090
  • ONLINE EXCLUSIVE PRICE
    307.50 €
    200.15 € / 12 pairs
    Save 107.35 €
    34% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fortix™ Technology increases protection with additional durability in the thumb crotch region, and extends the glove's useful life.

  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Fortix™ Technology extends product's useful life
  • Increased protection in the thumb crotch region
  • Water-based polyurethane enhances comfort and dexterity
  • Oeko-Tek™ 100 Certified
  • Solvent-free
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 5
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material Intercept Technology Yarn
Product Type Cut-resistant Gloves
Size 9
EN388 Standard 4X42C
For Use With (Application) Automotive Assembly, Machinery, Equipment Operation, and Metal Fabrication
Glove Cuff Style Knit
Series 11-738
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.