missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-738 Intercept™ Cut-Resistant Gloves with Targeted Reinforcement
GREENER_CHOICE

Product Code. p-7215018
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16099651

Brand: Ansell 11738060

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Fortix™ Technology increases protection with additional durability in the thumb crotch region, and extends the glove's useful life.

  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Fortix™ Technology extends product's useful life
  • Increased protection in the thumb crotch region
  • Water-based polyurethane enhances comfort and dexterity
  • Oeko-Tek™ 100 Certified
  • Solvent-free
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 5
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material Intercept Technology Yarn
Product Type Cut-resistant Gloves
Size 6
EN388 Standard 4X42C
For Use With (Application) Automotive Assembly, Machinery, Equipment Operation, and Metal Fabrication
Glove Cuff Style Knit
Series 11-738
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.