missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-739 Polyurethane-Coated Extended Cuff Gloves with Intercept™ Technology

Product Code. 16669093 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16669093 11 12 pairs
16619093 6 12 pairs
16629093 7 12 pairs
16639093 8 12 pairs
16649093 9 12 pairs
16659093 10 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16669093 Supplier Ansell Supplier No. 11739110
  • ONLINE EXCLUSIVE PRICE
    346.50 €
    226.79 € / 12 pairs
    Save 119.71 €
    34% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves combine comfort and dexterity with Intercept™ Technology to provide a high level of cut resistance. Polyurethane coating enhances grip capabilities in dry, as well as slightly oily environments.

  • A combination of comfort, sure handling, and cut resistance
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Ideal for safe use in controlling, and handling/assembly applications that involve contact with sharp pieces
  • Designed for use in dry as well as slightly oily environments
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) CE 0493, EN 407:2004, EN 420:2003 + A1:2009
Coating Material Polyurethane
Color Gray
Cut Resistance Rating Level D
Hand Specific
Latex Free No
Material Intercept Technology Yarn
Product Type Cut-resistant Gloves
Size 11
EN388 Standard 4X43D
For Use With (Application) Assembly, Metal Parts Handling, and Press Injection Molding
Glove Cuff Style Knit
Series 11-739
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.