missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-819 Nylon/Spandex ESD Gloves with FORTIX™ Nitrile Foam Coating
GREENER_CHOICE

Product Code. 17270157
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17270157 6 12 pairs
17280157 7 12 pairs
17290157 8 12 pairs
17200167 9 12 pairs
17210167 10 12 pairs
17220167 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17270157 Supplier Ansell Supplier No. 11819060
Add to basket
Add to basket
This item is not returnable. View return policy

Safety gloves with ESD functionality certified to EN 16350:2014, protecting products against electrostatic discharge. Ultra-thin FORTIX™ coating enhances grip, while also maintaining bare hand sensitivity for tasks that require dexterity and precision.

  • ESD functionality certified to EN 16350:2014
  • Can be used in explosion hazard zones as part of a grounding chain
  • Seamless, 18 gauge nylon and spandex liner helps maintain a bare-handed feel for tasks that require dexterity and precision
  • FORTIX™ nitrile foam coating enhances grip and provides long lasting protection
  • ZONZ™ Comfort Fit Technology ensures gloves are ultra breathable
  • Proprietary washing process helps to deliver cleaner, more comfortable gloves
  • Touchscreen functionality allows users to work with multiple touchscreen devices without glove removal
  • Dermatologically approved by Dermatest™
  • Certified by OEKO-TEX™
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 16350:2014, EN 420:2003 + A1:2009
Coated Area Palm
Accelerator Free Yes
Applications Industrial
Coating Material Nitrile Foam
Color Blue
Disposable Reusable
Exterior Finish Rough
Hand Specific
Laboratory/Cleanroom Use No
Latex Free Yes
Material Nylon, Spandex
Powdered Powder Free
Product Type ESD Gloves
Size 6
Sterility Non-sterile
EN388 Standard 3121A
For Use With (Application) Aerospace, Automotive, Logistics, and Machinery/Equipment
Glove Cuff Style Knit
Palm Type Coated
Series HyFlex™ 11-819
Yarn Gauge 18
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.