missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-731 Seamless Light Duty Liner Gloves, INTERCEPT™ Technology
GREENER_CHOICE

Product Code. 16875180
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16875180 6 12 pairs
16885180 7 12 pairs
16895180 8 12 pairs
16805190 9 12 pairs
16815190 10 12 pairs
16825190 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16875180 Supplier Ansell Supplier No. 11731060
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Light Duty 18 gauge seamless liners provide enhanced flexibility and tactility.

  • Provides enhanced range of motion and fingertip sensitivity
  • Thin PU coating provides high dexterity with high abrasion resistance (EN level 4)
  • Coating color masks and hides dirt
  • Also available in vendor pack
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) CE, EN 420:2003 + A1:2009; Category II
Construction Knit
Coating Material Polyurethane
Color Gray, White
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free No
Material HPPE, Fiberglass, Nylon, Spandex, Polyurethane
Product Type Cut-resistant Gloves
Size 6
EN388 Standard EN 388:4X42B, EN 388:4342
For Use With (Application) Assembly, inspection, press injection molding, fastening components, applying finish to materials, handling incoming goods
Glove Cuff Color Gray
Glove Cuff Style Knit
Liner Material Fiberglass, HPPE, Nylon, Spandex
Model 11731060
Quantity 12 Pairs/Bag, 12 Bags/Cs.
Series HyFlex™ 11-731
Length (English) 8.3 in.
Length (Metric) 21.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.