missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ HyFlex™ Sleeve with Intercept™ Technology

Product Code. 16825410
Change view
Click to view available options
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
16825410 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16825410 Supplier Ansell™ Supplier No. 11250120N
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ansell™ HyFlex™ Sleeve with Intercept™ Technology features a knitted bicep cuff to keep the sleeve in place

  • Available in different lengths and widths for optimal fit and comfort to fit.
  • Color-coding helps for easy sorting after laundry
  • Made of black yarn
  • Silicone-free for friendly paint and finish process
  • Compliant with FDA food handling regulations 21 CFR 177.1520 and 21 CFR 177.2600
  • Applications:
    • Press shop operations
    • Handling metal sheets and panels
    • Handling parts with sharp, rough edges
    • Handling structural and body components
    • Inspection, selecting, checking parts
    • Glass repair and replacement
    • Seizing structural and body parts
    • Body surface checking, marking and touching up
    • Adjusting systems and screwing
    • Adjusting panels parts components
TRUSTED_SUSTAINABILITY

Specifications

Size Narrow
Construction Knitted
Product Type Cut-resistant Sleeve
Material HPPE Yarn
Sterility Non-sterile
Style Ambidextrous
Disposable Reusable
Closure Type Elastic Upper with Knit Cuff
Seam Type Seamless
Cuff Style Sleeve
Model HyFlex™ 11-250
Certifications/Compliance FDA CFR 177.1520 and 21 CFR 177.2600; EN420:2003 + A1:2009; Category ll
Length (English) 12 in.
Length (Metric) 30 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.