Learn More
HOPEN Hygiene PP Gown with Collar, White
Product Code. 17236760 Shop All Medicom Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17236760 X-Large Pack of 50
17216760 Medium Pack of 50
17226760 Large Pack of 50
17246760 2X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17236760 Supplier HOPEN Supplier No. 40.632/04
  • ONLINE EXCLUSIVE PRICE
    76.00 €
    57.76 € / Pack of 50
    Save 18.24 €
    24% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Hygiene gown with zip and left inside pocket.
  • Hygiene PP Gown
  • Classic with collar
  • Elasticated wrists
  • With one interior pocket on the left side
  • With zip fastener
  • 40 g/m2
  • White
Specifications
Size X-Large
Material Polypropylene
Color White
Cuff Style Elastic
Includes Collar