missing translation for 'onlineSavingsMsg'
Learn More

Hygromycin B, MedChemExpress

Product Code. 16452199
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
1 g
5 g
Unit Size:
1g
200mg
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452199 1 g 1g
16462156 500 mg 500mg
16451426 200 mg 200mg
16410958 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16452199 Supplier Medchem Express Supplier No. HYB0490
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Hygromycin B is an aminoglycoside antibiotic active against prokaryotic and eukaryotic cells.

Hygromycin B is an aminoglycoside antibiotic active against prokaryotic and eukaryotic cells.

Specifications

Chemical Name or Material Hygromycin B
CAS 31282-04-9
Color White
Purity Grade Notes Research
Molecular Formula C20H37N3O13
Quantity 1 g
Synonym Hygrovetine
Solubility Information DMSO : 100 mg/mL (189.57 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 50 mg/mL (94.78 mM; Need ultrasonic)
SMILES O[C@H]([C@H]1O)C2(O[C@@]([C@@H](N)CO)([H])[C@@H]1O)O[C@@]([C@@H](O[C@@H]3CO)O[C@@]([C@H]([C@H](N)C4)O)([H])[C@@H]([C@H]4NC)O)([H])[C@@]([C@H]3O)([H])O2
Molecular Weight (g/mol) 527.52
Formula Weight 527.52
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.