Learn More
Ansell™ Hylite™ 47-409 Series Blue Nitrile Medium Weight Gloves
Product Code. 12630912
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
Product Code. 12630912
Brand: Ansell™ 47409070
Only null left
Add to basket
Add to basket
Versatile alternative to cotton, light leather and PVC. Special nitrile formulation provides an excellent combination of flexibility, mechanical resistance and oil repellency.
Ideal for assembly, inspection, light fabrication, materials handling, shipping and receiving, maintenance, and sheet metal work.
  • Natural hand shape - easier to wear for long periods.
  • Silicone-free.
  • Ensures cleanliness of metal sheets: important in steel and motor industries.
  • Cotton interlock lining with knitwrist for extra comfort
  • Nitrile-coated palm for exceptional cut
  • Offer puncture and abrasion resistance
  • Provide flexibility and dexterity
  • Machine washable; can be laundered repeatedly without becoming stiff and uncomfortable
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Certifications/Compliance CE Mark, ANSI, CATEGORY II EN 388 : 3111
Cuff Material Nitrile
Coating Material Nitrile
Color Green
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Mediumweight Gloves
Size 7
Glove Cuff Style Gold safety
Liner Material Interlock Cotton
Lining Lined
Palm Type Fully coated-palm
Quantity 144 Pairs
Resistant To Snag, puncture, abrasion and cut resistance
Series 47-409