missing translation for 'onlineSavingsMsg'
Learn More

Ansell Hynit™ 32-105 Light Duty Nitrile Coated Liquid-Repellent Gloves

Product Code. 12670822
Click to view available options
Size:
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 12670822

Brand: Ansell 32105100

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Fully impregnated with an unique liquid repellent nitrile formulation to provide durability in abrasive environments, while also protecting workers against oil, grease, and dirt.

  • Special nitrile formulation combines flexibility with abrasion resistance
  • Liquid-repellent impregnation repels oil, grease, and dirt
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 10
For Use With (Application) Automotive, Machinery/Equipment, Metal Fabrication, and Mining
Glove Cuff Style Open
Liner Material Cotton
Series 32-105
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.