Learn More
Ansell™ Hynit™ 32-815 Series Blue Nitrile Light Weight Gloves
Product Code. 12630842
Click to view available options
Size:
7
7.5
8
9
10
Unit Size:
12 pairs
Product Code. 12630842
Brand: Ansell™ 32815070
Only null left
Add to basket
Add to basket
Special nitrile formulation unites excellent resistance to abrasion with remarkable flexibility. Liquid-repellent impregnation repels oil, grease and dirt.
  • Cut and sewn construction
  • Nitrile coated
  • Polyester cotton liner
  • Blue and white color
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Certifications/Compliance CE Mark, ANSI, EN 388 : 3111
Cuff Material Nitrile
Coating Material Nitrile
Color Green
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Polycotton
Product Type Lightweight Gloves
Size 7
Glove Cuff Style Safety
Liner Material Interlock Knit Polyester Cotton
Lining Knit Lining
Palm Type Coated-palm
Quantity 144 Pairs
Resistant To Excellent snag, puncture, abrasion and cut resistance
Series 32-815