missing translation for 'onlineSavingsMsg'
Learn More

Hyoscine Hydrobromide Trihydrate, TRC

Product Code. 30134808
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134808 1 g 1g
30134142 500 mg 500mg
18070020 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134808 Supplier Toronto Research Chemicals Supplier No. TRCH6743081G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6533-68-2
Molecular Formula C17 H21 N O4 . Br H . 3 H2 O
Molecular Weight (g/mol) 438.31
Synonym Hyoscine Hydrobromide Trihydrate, Hyoscyamine Sulphate Imp. D (EP) as Hydrobromide Trihydrate, Hyoscine Butylbromide Imp. A (EP) as Hydrobromide Trihydrate, Atropine Imp. F (EP) as Hydrobromide Trihydrate, (αS)-α-(Hydroxymethyl)benzeneacetic acid (1α,2β,4β,5α,7β)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02, ́]non-7-yl ester hydrobromide hydrate (1:1:3), (αS)-α-(Hydroxymethyl)benzeneacetic acid (1α,2β,4β,5α,7β)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02, ́]non-7-yl ester hydrobromide trihydrate, α-(Hydroxymethyl)benzeneacetic acid [7(S)-(1α,2β,4β,5α,7β)]-9-methyl-3-oxa-9-azatricyclo[3.3.1.02, ́]non-7-yl ester hydrobromide trihydrate, 3-Oxa-9-azatricyclo[3.3.1.02, ́]nonane benzeneacetic acid deriv., (-)-Scopolamine hydrobromide trihydrate, Hyoscine hydrobromide trihydrate, Scopolamine hydrobromide trihydrate
IUPAC Name [(1R,2R,4S,5S)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-yl] (2S)-3-hydroxy-2-phenylpropanoate;trihydrate;hydrobromide
SMILES O.O.O.Br.CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H21NO4.BrH.3H2O/c1-18-13-7-11(8-14(18)16-15(13)22-16)21-17(20)12(9-19)10-5-3-2-4-6-10;;;;/h2-6,11-16,19H,7-9H2,1H3;1H;3*1H2/t11-,12-,13-,14+,15-,16+;;;;/m1..../s1
Formula Weight 437.1049
Percent Purity >95%
Chemical Name or Material Hyoscine Hydrobromide Trihydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.