Learn More
Safety First Aid HypaClens Emergency Eye Wash Dispenser, 10 x 20 mL Pods
Product Code. 31759396
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Each
Product Code. Sterility Unit Size
31759396 Sterile Each
1 options
Product Code. 31759396 Supplier Safety First Aid Supplier No. E4201
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
HypaClens Sterile Eyewash Pods gently wash the eye and provide sensitive and moisture replenishing relief for eyes that have been contaminated with particles or liquid.
  • HypaClens Sterile Eyewash Pods contain 0.9% sterile saline solution that is ideal for irrigating eyes and general wound washing applications.
  • The station can be refilled when required.
Specifications
Product Type Eyewash Solution
Sterility Sterile
Volume (Metric) 10 x 20 mL