missing translation for 'onlineSavingsMsg'
Learn More

Hypaconitine, MedChemExpress

Product Code. 16494797
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494797 50 mg 50mg
16438928 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16494797 Supplier Medchem Express Supplier No. HYN0267

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hypaconitine, an active and highly toxic constituent derived from Aconitum species, is widely used to treat rheumatism.

Hypaconitine, an active and highly toxic constituent derived from Aconitum species, is widely used to treat rheumatism.

Specifications

Chemical Name or Material Hypaconitine
CAS 6900-87-4
Color White
Purity Grade Notes Research
Molecular Formula C33H45NO10
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (81.21 mM; Need ultrasonic)
SMILES CO[C@@H](CC1)C23[C@@](C[C@@]4(O)[C@@H]5OC(C6=CC=CC=C6)=O)([H])[C@@]5([H])[C@](OC(C)=O)([C@@H](O)[C@@H]4OC)C7C2N(C)C[C@@]1(COC)[C@@]3([H])[C@H]7OC
Molecular Weight (g/mol) 615.71
Formula Weight 615.71
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.